MilliporeSigma
  • Mechanism of actin polymerization revealed by cryo-EM structures of actin filaments with three different bound nucleotides
Mechanism of actin polymerization revealed by cryo-EM structures of actin filaments with three different bound nucleotides
Proceedings of the National Academy of Sciences of the USA (2019)
Chou SZ and Pollard TD