MilliporeSigma
  • Improved selective electrocatalytic oxidation of phenols by tyrosinase-based carbon paste electrode biosensor
Improved selective electrocatalytic oxidation of phenols by tyrosinase-based carbon paste electrode biosensor
Electrochimica Acta (2000)
Rogers KR, et al.