MilliporeSigma
  • Reprogramming human amniotic fluid stem cells to functional pluripotency by manipulation of culture conditions
Reprogramming human amniotic fluid stem cells to functional pluripotency by manipulation of culture conditions
National Protocol Exchange (2012)
Moschidou D and Guillot PV