MilliporeSigma
  • Interferon-gamma is a crucial activator of early host immune defense against Mycobacterium ulcerans infection in mice
Interferon-gamma is a crucial activator of early host immune defense against Mycobacterium ulcerans infection in mice
PLoS Neglected Tropical Diseases (2016)
Bieri R, et al.