MilliporeSigma
  • Purification, characterization and degradation performance of a novel dextranase from Penicillium cyclopium CICC-4022
Purification, characterization and degradation performance of a novel dextranase from Penicillium cyclopium CICC-4022
International Journal of Molecular Sciences (2019)
Huang R, et al.