MilliporeSigma
  • Effect of milk hydrolysates on inflammation markers and drug-induced transcriptional alterations in cell-based models
Effect of milk hydrolysates on inflammation markers and drug-induced transcriptional alterations in cell-based models
Journal of Animal Science (2012)
Nielsen D, et al.