MilliporeSigma
  • Dietary protein content affects evolution for body size, body fat and viability in Drosophila melanogaster
Dietary protein content affects evolution for body size, body fat and viability in Drosophila melanogaster
Biology Letters (2010)
Kristensen TN, et al.