MilliporeSigma
  • Chickadees faced with unpredictable food increase fat reserves but certain components of their immune function decline
Chickadees faced with unpredictable food increase fat reserves but certain components of their immune function decline
Physiological and Biochemical Zoology : Pbz (2017)
Cornelius EA, et al.