MilliporeSigma
  • Second case of HOIP deficiency expands clinical features and defines inflammatory transcriptome regulated by LUBAC
Second case of HOIP deficiency expands clinical features and defines inflammatory transcriptome regulated by LUBAC
Frontiers in Immunology (2019)
Oda H, et al.