MilliporeSigma
  • Bacterial artificial chromosomes (BACs)-on-Beads? as a diagnostic platform for the rapid aneuploidy screening of products of conception
Bacterial artificial chromosomes (BACs)-on-Beads? as a diagnostic platform for the rapid aneuploidy screening of products of conception
Molecular Medicine Reports (2013)
Sheath KL, et al.