MilliporeSigma
  • Azocasein substrate for determination of proteolytic activity: reexamining a traditional method using bromelain samples
Azocasein substrate for determination of proteolytic activity: reexamining a traditional method using bromelain samples
BioMed Research International (2016)
Coelho DF, et al.