MilliporeSigma
  • The stability of almond beta-glucosidase during combined high pressure-thermal processing: a kinetic study
The stability of almond beta-glucosidase during combined high pressure-thermal processing: a kinetic study
Applied Microbiology and Biotechnology (2013)
Terefe N, et al.