MilliporeSigma
  • Glucose biosensors based on the immobilization of copper oxide and glucose oxidase within a carbon paste matrix
Glucose biosensors based on the immobilization of copper oxide and glucose oxidase within a carbon paste matrix
Talanta (2005)
Luque GL, et al.