MilliporeSigma
  • Efficient propylphosphonic anhydride (T3P) mediated synthesis of benzothiazoles, benzoxazoles and benzimidazoles
Efficient propylphosphonic anhydride (T3P) mediated synthesis of benzothiazoles, benzoxazoles and benzimidazoles
Tetrahedron Letters (2012)
Wen X, et al.