MilliporeSigma
  • Propylphosphonic anhydride (T3P): A remarkably efficient reagent for the one-pot transformation of aromatic, heteroaromatic, and aliphatic aldehydes to nitriles
Propylphosphonic anhydride (T3P): A remarkably efficient reagent for the one-pot transformation of aromatic, heteroaromatic, and aliphatic aldehydes to nitriles
Synlett (2009)
Augustine JK, et al.