MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Systematic optimization and experimental validation of ternary simulated moving bed chromatography systems
Systematic optimization and experimental validation of ternary simulated moving bed chromatography systems
Journal of Chromatography A (2014)
Agrawal G, et al.