MilliporeSigma
  • A validated normal phase HPLC method for simultaneous determination of drotaverine hydrochloride and omeprazole in pharmaceutical formulation
A validated normal phase HPLC method for simultaneous determination of drotaverine hydrochloride and omeprazole in pharmaceutical formulation
Asian Journal of Pharmaceutical and Clinical Research (2010)
Topagi SK, et al.