MilliporeSigma
  • N-acetyl-L-tryptophan, a substance-P receptor antagonist attenuates aluminum-induced spatial memory deficit in rats
N-acetyl-L-tryptophan, a substance-P receptor antagonist attenuates aluminum-induced spatial memory deficit in rats
Toxicology Mechanisms and Methods (2018)
Fernandes J, et al.