MilliporeSigma
  • A novel precursor in preparation and characterization of nickel oxide nanoparticles via thermal decomposition approach
A novel precursor in preparation and characterization of nickel oxide nanoparticles via thermal decomposition approach
Journal of alloys and compounds (2010)
Salavati-Niasari M, et al.