MilliporeSigma
  • Determination of lactic, acetic, succinic, and citric acids in table olives by HPLC/UV
Determination of lactic, acetic, succinic, and citric acids in table olives by HPLC/UV
Journal of Liquid Chromatography and Related Technologies (2001)
Cunha SC, et al.