MilliporeSigma
  • The use of carbon paste electrode in the direct voltammetric determination of tryptophan in pharmaceutical formulations
The use of carbon paste electrode in the direct voltammetric determination of tryptophan in pharmaceutical formulations
Journal of Pharmaceutical and Biomedical Analysis (2002)
Fiorucci AR, et al.