MilliporeSigma
  • Thermally robust and blinking suppressed core/graded-shell CdSe/CdSe1- xSx/CdS `giant?multishell semiconductor nanocrystals
Thermally robust and blinking suppressed core/graded-shell CdSe/CdSe1- xSx/CdS `giant?multishell semiconductor nanocrystals
Nanotechnology (2013)
Lau PC, et al.