MilliporeSigma
  • The development of electrically conductive polycaprolactone fumarate-polypyrrole composite materials for nerve regeneration
The development of electrically conductive polycaprolactone fumarate-polypyrrole composite materials for nerve regeneration
Biomaterials (2010)
Runge MB, et al.