MilliporeSigma
  • Structure of the cystathionine gamma-synthase MetB from Mycobacterium ulcerans
Structure of the cystathionine gamma-synthase MetB from Mycobacterium ulcerans
Acta Crystallographica. Section F, Structural Biology Communications (2011)
Clifton MC, et al.