MilliporeSigma
  • A high-performance liquid chromatographic assay for determination of cisplatin in plasma, cancer cell, and tumor samples.
A high-performance liquid chromatographic assay for determination of cisplatin in plasma, cancer cell, and tumor samples.
Journal of Pharmacological and Toxicological Methods (2005)
Lopez-Flores A, et al.