MilliporeSigma
  • Solubilities of magnesium-l-ascorbate, calcium-l-ascorbate, magnesium-l-glutamate, magnesium-d-gluconate, calcium-d-gluconate, calcium-d-heptagluconate, l-aspartic acid, and 3-nitrobenzoic acid in water
Solubilities of magnesium-l-ascorbate, calcium-l-ascorbate, magnesium-l-glutamate, magnesium-d-gluconate, calcium-d-gluconate, calcium-d-heptagluconate, l-aspartic acid, and 3-nitrobenzoic acid in water
The Journal of Chemical Thermodynamics (2008)
Mishelevich A and Apelblat A