MilliporeSigma
  • The initial repair response of articular cartilage after mechanically induced damage
The initial repair response of articular cartilage after mechanically induced damage
Journal of Orthopaedic Research : Official Publication of the Orthopaedic Research Society (2017)
van Haaften EE, et al.