MilliporeSigma
  • beta-Nicotinamide adenine dinucleotide is an inhibitory neurotransmitter in visceral smooth muscle
beta-Nicotinamide adenine dinucleotide is an inhibitory neurotransmitter in visceral smooth muscle
Proceedings of the National Academy of Sciences of the USA (2007)
Mutafova-Yambolieva VN, et al.