MilliporeSigma
  • The zebrafish kidney mutant zeppelin reveals that brca2/fancd1 is essential for pronephros development
The zebrafish kidney mutant zeppelin reveals that brca2/fancd1 is essential for pronephros development
Developmental Biology (2017)
Kroeger Jr PT, et al.