MilliporeSigma
  • BMAL1 controls glucose uptake through paired-homeodomain transcription factor 4 in differentiated Caco-2 cells.
BMAL1 controls glucose uptake through paired-homeodomain transcription factor 4 in differentiated Caco-2 cells.
American journal of physiology. Cell physiology (2019-06-20)
Whitney Sussman, Matthew Stevenson, Cyrus Mowdawalla, Samantha Mota, Louis Ragolia, Xiaoyue Pan
ABSTRACT
The transcription factor aryl hydrocarbon receptor nuclear translocator-like protein-1 (BMAL1) is an essential regulator of the circadian clock, which controls the 24-h cycle of physiological processes such as nutrient absorption. To examine the role of BMAL1 in small intestinal glucose absorption, we used differentiated human colon adenocarcinoma cells (Caco-2 cells). Here, we show that BMAL1 regulates glucose uptake in differentiated Caco-2 cells and that this process is dependent on the glucose transporter sodium-glucose cotransporter 1 (SGLT1). Mechanistic studies show that BMAL1 regulates glucose uptake by controlling the transcription of SGLT1 involving paired-homeodomain transcription factor 4 (PAX4), a transcriptional repressor. This is supported by the observation that clustered regularly interspaced short palindromic repeats (CRISPR)-CRISPR-associated endonuclease Cas9 (Cas9) knockdown of PAX4 increases SGLT1 and glucose uptake. Chromatin immunoprecipitation (ChIP) and ChIP-quantitative PCR assays show that the knockdown or overexpression of BMAL1 decreases or increases the binding of PAX4 to the hepatocyte nuclear factor 1-α binding site of the SGLT1 promoter, respectively. These findings identify BMAL1 as a critical mediator of small intestine carbohydrate absorption and SGLT1.
Sigma-Aldrich
MISSION® esiRNA, targeting human ARNTL
Pricing and availability is not currently available.