MilliporeSigma
  • The critical role for TAK1 in trichloroethylene-induced contact hypersensitivity in vivo and in CD4+ T cell function alteration by trichloroethylene and its metabolites in vitro.
The critical role for TAK1 in trichloroethylene-induced contact hypersensitivity in vivo and in CD4+ T cell function alteration by trichloroethylene and its metabolites in vitro.
Toxicology and applied pharmacology (2019-08-11)
Yao Pan, Xiaohong Hou, Qinghe Meng, Xiaohua Yang, Lanqin Shang, Xuetao Wei, Weidong Hao
ABSTRACT
Occupational exposure to trichloroethylene (TCE) has been associated with severe, generalized contact hypersensitivity (CHS) skin disorder, which is considered a delayed-type hypersensitivity reaction mediated by antigen-specific T cells. Transforming growth factor-β activated kinase-1 (TAK1) is essential for regulating the development and effector function of T cells. We hypothesized that disrupting TAK1 activity might inhibit TCE-induced CHS response. In this study, a local lymph node assay was employed to build a CHS model induced by TCE combined with the inducible-TAK1 deletion system to study the effect of TAK1 on it. It was observed that TAK1 deficiency ameliorated the TCE-induced CHS response and was associated with defective T cell expansion and activation and IFN-γ production in vivo. Furthermore, we investigated the effects of TCE and its metabolites trichloroacetic acid (TCA) and dichloroacetic acid (DCA) on CD4+ T cell function and the effect of TAK1 on it in vitro. The results showed that TCE, TCA and DCA augmented the proliferation, activation and differentiation of CD4+ T cells through Jnk MAPK and NF-κB pathways. TAK1 deletion significantly attenuated these effects induced by TCE, TCA or DCA on CD4+ T cells. In conclusion, it is suggested that TAK1 plays a critical role both in TCE-induced CHS response in vivo and in TCE and its metabolite-induced CD4+ T cell activation in vitro. Local inhibition of TAK1 might offer a promising alternative feasible strategy for TCE-induced CHS.
Sigma-Aldrich
Dichloroacetic acid, ReagentPlus®, ≥99%