MilliporeSigma
  • 24R,25-dihydroxyvitamin D3 modulates tumorigenicity in breast cancer in an estrogen receptor-dependent manner.
24R,25-dihydroxyvitamin D3 modulates tumorigenicity in breast cancer in an estrogen receptor-dependent manner.
Steroids (2019-07-16)
Anjali Verma, Zvi Schwartz, Barbara D Boyan
ABSTRACT
Vitamin D has long been prescribed as a supplement to breast cancer patients. This is partially motivated by data indicating that low serum vitamin D, measured as 25-hydroxyvitamin D3 [25(OH)D3], is associated with worsened cancer prognosis and decreased survival rates in cancer patients. However, clinical studies investigating the role of vitamin D supplementation in breast cancer treatment are largely inconclusive. One reason for this may be that many of these studies ignore the complexity of the vitamin D metabolome and the effects of these metabolites at the cellular level. Once ingested, vitamin D is metabolized into 37 different metabolites, including 25(OH)D3, which is the metabolite actually measured clinically, as well as 1,25(OH)2D3 and 24,25(OH)2D3. Recent work by our lab and others has demonstrated a role for 24R,25(OH)2D3, in the modulation of breast cancer tumors via an estrogen receptor α-dependent mechanism. This review highlights the importance of considering estrogen receptor status in vitamin d-associated prognostic studies of breast cancer and proposes a potential mechanism for 24R,25(OH)2D3 signaling in breast cancer cells.