MilliporeSigma
  • Effects of single and combined immunotherapy approach targeting amyloid β protein and α-synuclein in a dementia with Lewy bodies-like model.
Effects of single and combined immunotherapy approach targeting amyloid β protein and α-synuclein in a dementia with Lewy bodies-like model.
Alzheimer's & dementia : the journal of the Alzheimer's Association (2019-08-06)
Markus Mandler, Edward Rockenstein, Cassia Overk, Michael Mante, Jazmin Florio, Anthony Adame, Changyoun Kim, Radmila Santic, Achim Schneeberger, Frank Mattner, Sabine Schmidhuber, Gergana Galabova, Brian Spencer, Eliezer Masliah, Robert A Rissman
ABSTRACT
Immunotherapeutic approaches targeting amyloid β (Aβ) protein and tau in Alzheimer's disease and α-synuclein (α-syn) in Parkinson's disease are being developed for treating dementia with Lewy bodies. However, it is unknown if single or combined immunotherapies targeting Aβ and/or α-syn may be effective. Amyloid precursor protein/α-syn tg mice were immunized with AFFITOPEs® (AFF) peptides specific to Aβ (AD02) or α-syn (PD-AFF1) and the combination. AD02 more effectively reduced Aβ and pTau burden; however, the combination exhibited some additive effects. Both AD02 and PD-AFF1 effectively reduced α-syn, ameliorated degeneration of pyramidal neurons, and reduced neuroinflammation. PD-AFF1 more effectively ameliorated cholinergic and dopaminergic fiber loss; the combined immunization displayed additive effects. AD02 more effectively improved buried pellet test behavior, whereas PD-AFF1 more effectively improved horizontal beam test; the combined immunization displayed additive effects. Specific active immunotherapy targeting Aβ and/or α-syn may be of potential interest for the treatment of dementia with Lewy bodies.
Sigma-Aldrich
4-Maleimidobutyric acid N-hydroxysuccinimide ester, ≥98.0% (HPLC)