MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Acute and chronic toxicity of herbicides and their mixtures measured by Aliivibrio fischeri ecotoxicological assay.
Acute and chronic toxicity of herbicides and their mixtures measured by Aliivibrio fischeri ecotoxicological assay.
Ecotoxicology and environmental safety (2019-10-05)
Gergő Tóth, Judit Háhn, Balázs Kriszt, Sándor Szoboszlay
ABSTRACT
The purpose of our work was to determine the acute and chronic toxicity of three of the EU's most common herbicides - mesotrione, S-metolachlor, terbuthylazine - and their mixtures by Aliivibrio fischeri ecotoxicological assays. While comparing the sensitivity of the acute (30 min) Microtox® standard assay with the chronic (25 h) test adapted to microtiter plate, joint effects (antagonism, additive effect and synergism) to the bioluminescence inhibition (consequently the metabolic damage) in A. fischeri were also determined by Combination Index (CI) method. 30 min of exposure to mesotrione and S-metolachlor resulted in a relatively low acute toxicity (EC50 values were 118 and 265 mg/L), while terbuthylazine did not cause bioluminescence inhibition at all. Results showed that the chronic toxicity of S-metolachlor and terbuthylazine to A. fischeri (EC5010h = 59.2 and 4.9 mg/L and EC5015h = 54.0 and 9.6 mg/L, respectively) is larger by at least one order of magnitude than that after 30 min of contact time. Considering mesotrione no significant difference was experienced in toxicity. Regarding the EC50 values, all of the mixtures had synergistic joint effects in the acute assay. However, in the chronic test all the mixtures showed antagonistic responses with the exception of mesotrione and S-metolachlor (ratio 1:1) combination, which also had additive and synergistic effects after 10 and 15 h of exposure, similarly to the short-term test. This is also the first report of the joint effects of these herbicides. The chronic test is a more sensitive indicator to the active ingredients; both acute and chronic assays supply valuable data of the toxic properties of the pesticides. Moreover, the short- and long-term joint effects of their mixtures supporting a more accurate and reliable risk assessment.
Product Description
Supelco
Mesotrione, PESTANAL®, analytical standard
Supelco
Metolachlor, predominantly S-Metolachlor, PESTANAL®, analytical standard