MilliporeSigma
  • Synergistic Effects of Nobiletin and Sorafenib Combination on Metastatic Prostate Cancer Cells.
Synergistic Effects of Nobiletin and Sorafenib Combination on Metastatic Prostate Cancer Cells.
Nutrition and cancer (2019-05-01)
Gamze Guney Eskiler, Asuman Ozkan Deveci, Cemil Bilir, Suleyman Kaleli
ABSTRACT
Objective: Herein we, for the first time, investigated a potential synergistic effect of nobiletin (NOB) and sorafenib (SOR) on PC-3 prostate cancer and HUVEC control cell lines. Methods: In order to determine the cytotoxic and apoptotic effects of the combination of NOB and SOR, WST-1, Annexin V, and cell cycle analysis were performed. The potential molecular mechanism of apoptotic cell death was assessed by Bax, Bcl-2, CCDN1, Rb1, and CDKN1A gene expression and acridine orange (AO) and DAPI staining. Results: Our results indicated that NOB and SOR combination had a significant inhibitory effect on the viability of PC-3 cells with less toxicity on HUVEC cells than SOR alone (P < 0.01). NOB and SOR combination significantly caused much more apoptotic cell death and cell cycle arrest at G0/G1 phase by up-regulation of Bax, Rb1, and CDKN1A levels in PC-3 cells (P < 0.01). Therefore, strong synergistic effects between NOB and SOR were analyzed (CI < 1). Conclusion: NOB and SOR combination was more effective than SOR and NOB alone and reduced the exposure time for SOR and NOB in PC-3 cells. Combination strategy is a therapeutic potential to improve efficacy and reduce side-effect of SOR for the treatment of metastatic prostate cancer cells.
Sigma-Aldrich
Acridine Orange hydrochloride solution, 10 mg/mL in H2O, ≥95.0% (HPLC)
Sigma-Aldrich
Nitrosobenzene, ≥97%