MilliporeSigma
  • The modulating effect of lipid bilayer/p-coumaric acid interactions on electrical properties of model lipid membranes and human glioblastoma cells.
The modulating effect of lipid bilayer/p-coumaric acid interactions on electrical properties of model lipid membranes and human glioblastoma cells.
Bioorganic chemistry (2019-09-09)
Monika Naumowicz, Magdalena Kusaczuk, Marcin Andrzej Kruszewski, Miroslav Gál, Rafał Krętowski, Marzanna Cechowska-Pasko, Joanna Kotyńska
ABSTRACT
Biological membranes are one of the most important elements of living cells determining their permeability to the active compounds. Still, little is known about the drug-membrane interactions in terms of pharmacological properties of potential drugs. Chemoprevention based on natural compounds is becoming a strong trend in modern oncopharmacology, and p-coumaric acid (p-CoA) is one such compound with tentative anticancer activity. The microelectrophoretic mobility measurements and electrochemical impedance spectroscopy were applied to study the effects of p-CoA on electrical properties of liposomes, spherical bilayers, and human glioblastoma cell membranes. Our results demonstrated that after treatment with p-CoA, the surface charge of LBC3, LN-229 and LN-18 cell lines was significantly changed in alkaline pH solutions, but not in acidic pH solutions. In contrast, no changes in surface charge density values were registered for phosphatidylethanolamine liposomal membranes and A172 cell membranes after p-CoA treatment. The impedance data showed an increase in values of both the electrical capacitance and the electrical resistance, indicating that p-CoA can be partially inserted into the phosphatidylcholine bilayers. The MTT assay showed cell line-dependent cytotoxic effect of p-CoA. Further molecular analyses revealed the ATP depletion and gene transcription modulation, which might indicate organelle membrane-crossing potential of p-CoA. These results suggest, that changes in surface charge of membranes of living cells not only might be potential predictor of membrane permeability, but also indicate differential composition of cell membranes in various cell lines. Thus further multidirectional analyses are required to implement electrochemical methods as standard testing procedures during drug development process.
Sigma-Aldrich
p-Coumaric acid, ≥98.0% (HPLC)