MilliporeSigma
  • Energy storage studies of bare and doped vanadium pentoxide,(V 1.95 M 0.05) O 5, M= Nb, Ta, for lithium ion batteries
Energy storage studies of bare and doped vanadium pentoxide,(V 1.95 M 0.05) O 5, M= Nb, Ta, for lithium ion batteries
Energy & Environmental Science (2011)
Sakunthala A, et al.