MilliporeSigma
  • An Acid Protease Produced by Monilinia fructigena in vitro and in Infected Apple Fruits, and its Possible Role in Pathogenesis
An Acid Protease Produced by Monilinia fructigena in vitro and in Infected Apple Fruits, and its Possible Role in Pathogenesis
Microbiology (1982)
Hislop EC, et al.