MilliporeSigma
  • Role of apoptosis in erosive and reticular oral lichen planus exhibiting variable epithelial thickness
Role of apoptosis in erosive and reticular oral lichen planus exhibiting variable epithelial thickness
Brazilian Dental Journal (2008)
Brant JMC, et al.