MilliporeSigma
  • Structure of beta-galactosidase at 3.2-AA resolution obtained by cryo-electron microscopy
Structure of beta-galactosidase at 3.2-AA resolution obtained by cryo-electron microscopy
Proceedings of the National Academy of Sciences of the USA (2014)
Bartesaghi A, et al.