MilliporeSigma
  • In vitro Nitrate Reductase Activity Assay from Arabidopsis Crude Extracts
In vitro Nitrate Reductase Activity Assay from Arabidopsis Crude Extracts
Bio-protocol (2018)
Kim JY and Seo HS