MilliporeSigma
  • Antioxidant properties of propofol and erythropoietin after closed head injury in rats
Antioxidant properties of propofol and erythropoietin after closed head injury in rats
Progress in Neuro-Psychopharmacology & Biological Psychiatry (2005)
Ozturk E, et al.