MilliporeSigma
  • Polycationic β-cyclodextrin ?click clusters?: monodisperse and versatile scaffolds for nucleic acid delivery
Polycationic β-cyclodextrin ?click clusters?: monodisperse and versatile scaffolds for nucleic acid delivery
Journal of the American Chemical Society (2008)
Srinivasachari S, et al.