MilliporeSigma
  • Pendant polymer: amino-β-cyclodextrin: siRNA guest: host nanoparticles as efficient vectors for gene silencing
Pendant polymer: amino-β-cyclodextrin: siRNA guest: host nanoparticles as efficient vectors for gene silencing
Journal of the American Chemical Society (2012)
Kulkarni A, et al.