MilliporeSigma
  • Proteomics in the liver of gilthead sea bream (Sparus aurata) to elucidate the cellular response induced by the intake of maslinic acid.
Proteomics in the liver of gilthead sea bream (Sparus aurata) to elucidate the cellular response induced by the intake of maslinic acid.
Proteomics (2011-07-14)
Eva Rufino-Palomares, Fernando J Reyes-Zurita, Carlos A Fuentes-Almagro, Manuel de la Higuera, José A Lupiáñez, Juan Peragón
ABSTRACT
Maslinic acid (MA) is a pentacyclic triterpene used as a feed additive to stimulate growth, protein-turnover rates, and hyperplasia in fish. To further our understanding of cellular mechanisms underlying the action of MA, we have used 2-DE coupled with MS to identify proteins differentially expressed in the livers of juvenile gilthead sea bream (Sparus aurata) grown under fish-farm conditions and fed with a 100 mg/kg MA-enriched diet (MA(100)). After the comparison of the protein profiles from MA(100) fed fish and from control, 49 protein spots were found to be altered in abundance (≥2-fold). Analysis by MALDI-TOF/TOF allowed the unambiguous identification of 29 spots, corresponding to 19 different proteins. These proteins were: phosphoglucomutase, phosphoglucose isomerase, S-adenosyl methionine-dependent methyltransferase class I, aldehyde dehydrogenase, catalase, 6-phosphogluconate dehydrogenase, fumarylacetoacetate hydrolase, 4-hydroxyphenylpyruvic dioxygenase, methylmalonate-semialdehyde dehydrogenase, lysozyme, urate oxidase, elongation factor 2, 60 kDa heat-shock protein, 58 kDa glucose-regulated protein, cytokeratin E7, type-II keratin, intermediate filament proteins, 17-β-hydroxysteroid dehydrogenase type 4, and kinase suppressor of Ras1. Western blot analysis of kinase suppressor of Ras1, glucose 6-phosphate dehydrogenase, elongation factor 2, 60 kDa heat-shock protein, and catalase supported the proteome evidence. Based on the changes found in the protein-expression levels of these proteins, we proposed a cellular-signalling pathway to explain the hepatic-cell response to the intake of a diet containing MA.
Sigma-Aldrich
Maslinic acid, ≥98% (HPLC)