MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Small molecule drug screening in Drosophila identifies the 5HT2A receptor as a feeding modulation target.
Small molecule drug screening in Drosophila identifies the 5HT2A receptor as a feeding modulation target.
Scientific reports (2013-07-03)
Gabriel Gasque, Stephen Conway, Juan Huang, Yi Rao, Leslie B Vosshall
ABSTRACT
Dysregulation of eating behavior can lead to obesity, which affects 10% of the adult population worldwide and accounts for nearly 3 million deaths every year. Despite this burden on society, we currently lack effective pharmacological treatment options to regulate appetite. We used Drosophila melanogaster larvae to develop a high-throughput whole organism screen for drugs that modulate food intake. In a screen of 3630 small molecules, we identified the serotonin (5-hydroxytryptamine or 5-HT) receptor antagonist metitepine as a potent anorectic drug. Using cell-based assays we show that metitepine is an antagonist of all five Drosophila 5-HT receptors. We screened fly mutants for each of these receptors and found that serotonin receptor 5-HT2A is the sole molecular target for feeding inhibition by metitepine. These results highlight the conservation of molecular mechanisms controlling appetite and provide a method for unbiased whole-organism drug screens to identify novel drugs and molecular pathways modulating food intake.
Sigma-Aldrich
Mifepristone, ≥98%