MilliporeSigma
  • The NF-kappa B signaling pathway is not required for Fas ligand gene induction but mediates protection from activation-induced cell death.
The NF-kappa B signaling pathway is not required for Fas ligand gene induction but mediates protection from activation-induced cell death.
The Journal of biological chemistry (2000-06-06)
I Rivera-Walsh, M E Cvijic, G Xiao, S C Sun
ABSTRACT
Stimulation of T cells by antigens or mitogens triggers multiple signaling pathways leading to activation of genes encoding interleukin-2 and other growth-regulatory cytokines. The same stimuli also activate the gene encoding an apoptosis-inducing molecule, Fas ligand (FasL), which contributes to activation-induced cell death. It has been proposed that the signaling pathways involved in cytokine gene induction also contribute to activation-induced FasL expression; however, genetic evidence for this proposal is lacking. In the present study, the role of the NF-kappaB signaling pathway in FasL gene expression was examined using a mutant T cell line deficient in an essential NF-kappaB signaling component, IkappaB kinase gamma. These mutant cells have a blockade in signal-induced activation of NF-kappaB but remained normal in the activation of NF-AT and AP-1 transcription factors. Interestingly, the NF-kappaB signaling defect has no effect on mitogen-stimulated FasL gene expression, although it completely blocks the interleukin-2 gene induction. We further demonstrate that NF-kappaB activation is required for protecting T cells from apoptosis induction by mitogens and an agonistic anti-Fas antibody. These genetic results suggest that the NF-kappaB signaling pathway is not required for activation-induced FasL expression but rather mediates cell growth and protection from activation-induced cell death.
Sigma-Aldrich
Anti-Fas Antibody (human, activating), clone CH11, clone CH11, Upstate®, from mouse