MilliporeSigma
  • Disruption of NSD1 in Head and Neck Cancer Promotes Favorable Chemotherapeutic Responses Linked to Hypomethylation.
Disruption of NSD1 in Head and Neck Cancer Promotes Favorable Chemotherapeutic Responses Linked to Hypomethylation.
Molecular cancer therapeutics (2018-04-11)
Nam Bui, Justin K Huang, Ana Bojorquez-Gomez, Katherine Licon, Kyle S Sanchez, Sean N Tang, Alex N Beckett, Tina Wang, Wei Zhang, John Paul Shen, Jason F Kreisberg, Trey Ideker
ABSTRACT
Human papillomavirus (HPV)-negative head and neck squamous cell carcinoma (HNSCC) represents a distinct classification of cancer with worse expected outcomes. Of the 11 genes recurrently mutated in HNSCC, we identify a singular and substantial survival advantage for mutations in the gene encoding Nuclear Set Domain Containing Protein 1 (NSD1), a histone methyltransferase altered in approximately 10% of patients. This effect, a 55% decrease in risk of death in NSD1-mutated versus non-mutated patients, can be validated in an independent cohort. NSD1 alterations are strongly associated with widespread genome hypomethylation in the same tumors, to a degree not observed for any other mutated gene. To address whether NSD1 plays a causal role in these associations, we use CRISPR-Cas9 to disrupt NSD1 in HNSCC cell lines and find that this leads to substantial CpG hypomethylation and sensitivity to cisplatin, a standard chemotherapy in head and neck cancer, with a 40% to 50% decrease in the IC50 value. Such results are reinforced by a survey of 1,001 cancer cell lines, in which loss-of-function NSD1 mutations have an average 23% decrease in cisplatin IC50 value compared with cell lines with wild-type NSD1Significance: This study identifies a favorable subtype of HPV-negative HNSCC linked to NSD1 mutation, hypomethylation, and cisplatin sensitivity. Mol Cancer Ther; 17(7); 1585-94. ©2018 AACR.