MilliporeSigma
  • Chemotherapeutic agent 5-fluorouracil increases survival of SOD1 mouse model of ALS.
Chemotherapeutic agent 5-fluorouracil increases survival of SOD1 mouse model of ALS.
PloS one (2019-01-15)
Amaya Rando, Miriam de la Torre, Anna Martinez-Muriana, Pilar Zaragoza, Antonio Musaro, Sara Hernández, Xavier Navarro, Janne M Toivonen, Rosario Osta
ABSTRACT
Amyotrophic lateral sclerosis (ALS) is a lethal motor neuron disease with no cure. Currently there are only two ALS drugs approved by the FDA, both with a limited therapeutic effect. In the search for drug candidates for ALS, we studied the effect of known stem cell mobilizing agents (treatment) and antimetabolite 5-fluorouracil (5-FU) (anti-treatment) in SOD1G93A model of ALS. Surprisingly, we found that anti-cancer drug 5-FU increases lifespan, delays the disease onset and improves motor performance in ALS mice. Although we were not able to demonstrate the mechanistic basis of the beneficial 5-FU action in ALS mice, our findings suggest that 5-FU or similar drugs are possible drug candidates for the treatment of motor neuron diseases through drug repurposing.
Sigma-Aldrich
Anti-SOD1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution