MilliporeSigma
  • Application of thiopropyl sepharose 6B for removal of PCR inhibitors from DNA extracts of a thigh bone recovered from the sea.
Application of thiopropyl sepharose 6B for removal of PCR inhibitors from DNA extracts of a thigh bone recovered from the sea.
International journal of legal medicine (2003-06-24)
Erik Sørensen, Steen Holger Hansen, Birthe Eriksen, Niels Morling
ABSTRACT
PCR amplification of DNA from forensic samples often proves difficult due to the presence of inhibitors of the polymerase chain reaction. One possible way to remove PCR inhibitors from a DNA extract is the use of the affinity resin thiopropyl sepharose 6B (TS), which has been used previously for the removal of PCR inhibitors in DNA extracts originating from stains on clothing. Here we show that TS is efficient also for the removal of inhibitors from PCR extracts from a highly decomposed human thigh bone. TS treatment, however, leads to a substantial loss of DNA making the technique best suited when substantial amounts of DNA are present.